missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dihydroxy-5-nitrobenzaldehyde 98.0+%, TCI America™

Catalog No. D364525G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D364525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D364525G Supplier TCI America Supplier No. D364525G

Chemical Identifiers

CAS 116313-85-0
Molecular Formula C7H5NO5
Molecular Weight (g/mol) 183.119
MDL Number MFCD00871897
InChI Key BBFJODMCHICIAA-UHFFFAOYSA-N
PubChem CID 5748957
SMILES C1=C(C=C(C(=C1[N+](=O)[O-])O)O)C=O

Specifications

Melting Point 147°C
Color Yellow
Quantity 25 g
Formula Weight 183.12
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,4-Dihydroxy-5-nitrobenzaldehyde
EINECSNumber (3)-4486
RTECSNumber CU5665000
TSCA No