missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,4-Dihydro-2H-pyrido[1,2-a]pyrimidin-2-one 98.0+%, TCI America™
Supplier: TCI America D139325G
Specifications
| 3,4-Dihydro-2H-pyrido[1,2-a]pyrimidin-2-one | |
| White-Yellow | |
| MFCD00042798 | |
| Acid Captor H | |
| C1CN2C=CC=CC2=NC1=O | |
| 148.165 | |
| 148.17 | |
| Crystalline Powder |
| 5439-14-5 | |
| C8H8N2O | |
| 25 g | |
| XQIOBBHIEUGFCI-UHFFFAOYSA-N | |
| 3,4-dihydropyrido[1,2-a]pyrimidin-2-one | |
| 79500 | |
| ≥98.0% (HPLC,T) |
Chemical Identifiers
| 5439-14-5 | |
| 148.165 | |
| XQIOBBHIEUGFCI-UHFFFAOYSA-N | |
| 79500 | |
| C1CN2C=CC=CC2=NC1=O |
| C8H8N2O | |
| MFCD00042798 | |
| Acid Captor H | |
| 3,4-dihydropyrido[1,2-a]pyrimidin-2-one |
Safety and Handling
TSCA : No