missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dihydro-2H-pyrido[1,2-a]pyrimidin-2-one 98.0+%, TCI America™

Catalog No. p-7126709
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D13935G 5 g
D139325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D13935G Supplier TCI America Supplier No. D13935G
Only null left

Chemical Identifiers

CAS 5439-14-5
Molecular Formula C8H8N2O
Molecular Weight (g/mol) 148.165
MDL Number MFCD00042798
InChI Key XQIOBBHIEUGFCI-UHFFFAOYSA-N
Synonym Acid Captor H
PubChem CID 79500
IUPAC Name 3,4-dihydropyrido[1,2-a]pyrimidin-2-one
SMILES C1CN2C=CC=CC2=NC1=O

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 148.17
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3,4-Dihydro-2H-pyrido[1,2-a]pyrimidin-2-one