accessibility menu, dialog, popup

UserName

CAS RN 5439-14-5

IUPAC Name: 2H,3H,4H-pyrido[1,2-a]pyrimidin-2-one
Synonyms: 3H,4H-pyrido[1,2-a]pyrimidin-2-one
Molecular Weight (g/mol): 148.17
Molecular Formula: C8H8N2O
InChi Key: XQIOBBHIEUGFCI-UHFFFAOYSA-N
SMILES: O=C1CCN2C=CC=CC2=N1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results