missing translation for 'onlineSavingsMsg'
Learn More

3,4-Difluorotoluene 98.0+%, TCI America™

Catalog No. D40541G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D40541G 1 g
D40545G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D40541G Supplier TCI America Supplier No. D40541G
Only null left

Chemical Identifiers

CAS 2927-34-6
Molecular Formula C7H6F2
Molecular Weight (g/mol) 128.12
MDL Number MFCD00075087
InChI Key FZMPLKVGINKUJZ-UHFFFAOYSA-N
PubChem CID 2733400
IUPAC Name 1,2-difluoro-4-methylbenzene
SMILES CC1=CC=C(F)C(F)=C1

Specifications

Color Yellow
Boiling Point 112°C
Quantity 1 g
UN Number 1993
Synonym 3,4-difluorotoluene, benzene, 1,2-difluoro-4-methyl, 1,2-difluoro-4-methyl-benzene, 3,4-difluoro methyl benzene, 3,4 difluorotoluene, pubchem1615, 3, 4-difluorotoluene, 3,4-difluoro toluene
Formula Weight 128.12
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3,4-Difluorotoluene