CAS RN 2927-34-6
CAS RN 2927-34-6
IUPAC Name:
1,2-difluoro-4-methylbenzene
Synonyms:
1,2-difluoro-4-methylbenzene
Molecular Weight (g/mol):
128.12
Molecular Formula:
C7H6F2
InChi Key:
FZMPLKVGINKUJZ-UHFFFAOYSA-N
SMILES:
CC1=CC=C(F)C(F)=C1
1
–
2
of
2
results
1
3,4-Difluorotoluene, 98%
CAS: 2927-34-6 Molecular Formula: C7H6F2 Molecular Weight (g/mol): 128.12 MDL Number: MFCD00075087 InChI Key: FZMPLKVGINKUJZ-UHFFFAOYSA-N PubChem CID: 2733400 IUPAC Name: 1,2-difluoro-4-methylbenzene SMILES: CC1=CC=C(F)C(F)=C1
2
3,4-Difluorotoluene 98.0+%, TCI America™
CAS: 2927-34-6 Molecular Formula: C7H6F2 Molecular Weight (g/mol): 128.12 MDL Number: MFCD00075087 InChI Key: FZMPLKVGINKUJZ-UHFFFAOYSA-N PubChem CID: 2733400 IUPAC Name: 1,2-difluoro-4-methylbenzene SMILES: CC1=CC=C(F)C(F)=C1