missing translation for 'onlineSavingsMsg'
Learn More

3,4-Difluorotoluene, 98%

Catalog No. p-7051692
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4178903 1 g
AA4178906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4178903 Supplier Thermo Scientific Chemicals Supplier No. 04178903
Only null left

CAS: 2927-34-6 | C7H6F2 | 128.12 g/mol

Used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2927-34-6
Molecular Formula C7H6F2
Molecular Weight (g/mol) 128.12
MDL Number MFCD00075087
InChI Key FZMPLKVGINKUJZ-UHFFFAOYSA-N
PubChem CID 2733400
IUPAC Name 1,2-difluoro-4-methylbenzene
SMILES CC1=CC=C(F)C(F)=C1

Specifications

Density 1.120 g/mL
Boiling Point 110°C to 113°C
Flash Point 25°C (77°F)
Odor Aromatic
Refractive Index 1.45
Quantity 1 g
UN Number UN1993
Synonym 3,4-difluorotoluene, benzene, 1,2-difluoro-4-methyl, 1,2-difluoro-4-methyl-benzene, 3,4-difluoro methyl benzene, 3,4 difluorotoluene, pubchem1615, 3, 4-difluorotoluene, 3,4-difluoro toluene
Solubility Information Insoluble in water.
Formula Weight 128.12
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 3,4-Difluorotoluene
Show More Show Less
Hazard Category H226
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only