missing translation for 'onlineSavingsMsg'
Learn More

3,4-Difluorophenylglyoxal hydrate, 98%, dry wt. basis

Catalog No. AAL2027103
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL2027103 1 g
AAL2027106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL2027103 Supplier Thermo Scientific Chemicals Supplier No. L2027103
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 79784-34-2 | C8H4F2O2 | 170.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 79784-34-2
Molecular Formula C8H4F2O2
Molecular Weight (g/mol) 170.12
MDL Number MFCD01733155
InChI Key VZRYIZGMQICGSF-UHFFFAOYSA-N
Synonym 3,4-difluorophenylglyoxal hydrate, 2-3,4-difluorophenyl-2-oxoacetaldehyde hydrate, pubchem12210, 3,4-difluorophenylglyoxal monohydrate, 3,4-difluorophenylglyoxalhydrate, benzeneacetaldehyde,3,4-difluoro-a-oxo, 2-3,4-difluorophenyl-2-oxoethanal, hydrate
PubChem CID 2782289
IUPAC Name 2-(3,4-difluorophenyl)-2-oxoacetaldehyde;hydrate
SMILES FC1=CC=C(C=C1F)C(=O)C=O

Specifications

Melting Point 85°C to 90°C
Quantity 1 g
Beilstein 2574671
Formula Weight 170.12 (Anhydrous)
Percent Purity 98%
Assay (dry wt.)
Chemical Name or Material 3,4-Difluorophenylglyoxal hydrate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
RTECSNumber CY1430000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only