missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,4-Difluorobenzenesulfonamide 98.0+%, TCI America™
$48.13 - $150.16
Chemical Identifiers
| CAS | 108966-71-8 |
|---|---|
| Molecular Formula | C6H5F2NO2S |
| Molecular Weight (g/mol) | 193.17 |
| MDL Number | MFCD00728800 |
| InChI Key | VFVVRYNJTGHAIE-UHFFFAOYSA-N |
| PubChem CID | 822865 |
| IUPAC Name | 3,4-difluorobenzene-1-sulfonamide |
| SMILES | NS(=O)(=O)C1=CC=C(F)C(F)=C1 |
Chemical Identifiers
| 108966-71-8 | |
| 193.17 | |
| VFVVRYNJTGHAIE-UHFFFAOYSA-N | |
| 3,4-difluorobenzene-1-sulfonamide |
| C6H5F2NO2S | |
| MFCD00728800 | |
| 822865 | |
| NS(=O)(=O)C1=CC=C(F)C(F)=C1 |
Specifications
| 108966-71-8 | |
| White | |
| MFCD00728800 | |
| 3,4-difluorobenzenesulphonamide, 3,4-difluorobenzene-1-sulfonamide, 3,4-difluoro-benzenesulfonamide, benzenesulfonamide, 3,4-difluoro, acmc-20ahuj, pubchem11788, benzenesulfonamide,3,4-difluoro, 3,4-difluorobenzenesulfonamide, 3,4-difluoro benzene sulfonamide, benzenesulfonamide, 3,4-difluoro-9ci | |
| NS(=O)(=O)C1=CC=C(F)C(F)=C1 | |
| 193.17 | |
| 193.17 | |
| Crystalline Powder |
| 88°C | |
| C6H5F2NO2S | |
| 1 g | |
| VFVVRYNJTGHAIE-UHFFFAOYSA-N | |
| 3,4-difluorobenzene-1-sulfonamide | |
| 822865 | |
| ≥98.0% (GC) | |
| 3,4-Difluorobenzenesulfonamide |
Safety and Handling
TSCA : No