missing translation for 'onlineSavingsMsg'
Learn More

3,4-Difluorobenzenesulfonamide 98.0+%, TCI America™

Catalog No. d48921g
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
5 g
Catalog No. Quantity
D48921G 1 g
D48925G 5 g
2 options

Catalog No. D48921G

Supplier: TCI America D48921G

Only null left

Chemical Identifiers

CAS 108966-71-8
Molecular Formula C6H5F2NO2S
Molecular Weight (g/mol) 193.17
MDL Number MFCD00728800
InChI Key VFVVRYNJTGHAIE-UHFFFAOYSA-N
PubChem CID 822865
IUPAC Name 3,4-difluorobenzene-1-sulfonamide
SMILES NS(=O)(=O)C1=CC=C(F)C(F)=C1

Specifications

Melting Point 88°C
Color White
Quantity 1 g
Synonym 3,4-difluorobenzenesulphonamide, 3,4-difluorobenzene-1-sulfonamide, 3,4-difluoro-benzenesulfonamide, benzenesulfonamide, 3,4-difluoro, acmc-20ahuj, pubchem11788, benzenesulfonamide,3,4-difluoro, 3,4-difluorobenzenesulfonamide, 3,4-difluoro benzene sulfonamide, benzenesulfonamide, 3,4-difluoro-9ci
Formula Weight 193.17
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,4-Difluorobenzenesulfonamide