missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,4-Difluorobenzaldehyde 97.0+%, TCI America™
Supplier: TCI America D234025G
Specifications
| 3,4-Difluorobenzaldehyde | |
| Yellow | |
| C7H4F2O | |
| 25 g | |
| JPHKMYXKNKLNDF-UHFFFAOYSA-N | |
| 3,4-difluorobenzaldehyde | |
| 588088 | |
| ≥97.0% (GC) |
| 34036-07-2 | |
| 180°C | |
| MFCD00010328 | |
| benzaldehyde, 3,4-difluoro, pubchem1413, 3,4 difluorobenzaldehyde, 3,4-diflourobenzaldehyde, 3.4-difluorobenzaldehyde, 3,4-difluoro-benzaldehye, 3,4-difluro-benzaldehyde, acmc-1ajc9, 3, 4-difluorobenzaldehyde, 3,4-difluoro benzaldehyde | |
| C1=CC(=C(C=C1C=O)F)F | |
| 142.105 | |
| 142.10 | |
| Liquid |
Chemical Identifiers
| 34036-07-2 | |
| 142.105 | |
| JPHKMYXKNKLNDF-UHFFFAOYSA-N | |
| 588088 | |
| C1=CC(=C(C=C1C=O)F)F |
| C7H4F2O | |
| MFCD00010328 | |
| benzaldehyde, 3,4-difluoro, pubchem1413, 3,4 difluorobenzaldehyde, 3,4-diflourobenzaldehyde, 3.4-difluorobenzaldehyde, 3,4-difluoro-benzaldehye, 3,4-difluro-benzaldehyde, acmc-1ajc9, 3, 4-difluorobenzaldehyde, 3,4-difluoro benzaldehyde | |
| 3,4-difluorobenzaldehyde |
Safety and Handling
TSCA : No