missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,4-Difluoroanisole 97.0+%, TCI America™
Supplier: TCI America D285625G
Specifications
| 3,4-Difluoroanisole | |
| Yellow | |
| MFCD00010730 | |
| 1993 | |
| DSEVNUCNUTYYHW-UHFFFAOYSA-N | |
| 1,2-difluoro-4-methoxybenzene | |
| 568250 | |
| ≥97.0% (GC) |
| 115144-40-6 | |
| C7H6F2O | |
| 25 g | |
| 3,4-difluoroanisole, 1,2-difluoro-4-methoxy-benzene, benzene, 1,2-difluoro-4-methoxy, 3,4-difluoro-1-methoxybenzene, 1,2-difluor-4-methoxybenzol, pubchem3457, 3,4-difluoro-anisole, difluorobenzene,1-methoxy, acmc-2099pe | |
| COC1=CC(=C(C=C1)F)F | |
| 144.121 | |
| 144.12 | |
| Liquid |
Chemical Identifiers
| 115144-40-6 | |
| 144.121 | |
| DSEVNUCNUTYYHW-UHFFFAOYSA-N | |
| 568250 | |
| COC1=CC(=C(C=C1)F)F |
| C7H6F2O | |
| MFCD00010730 | |
| 3,4-difluoroanisole, 1,2-difluoro-4-methoxy-benzene, benzene, 1,2-difluoro-4-methoxy, 3,4-difluoro-1-methoxybenzene, 1,2-difluor-4-methoxybenzol, pubchem3457, 3,4-difluoro-anisole, difluorobenzene,1-methoxy, acmc-2099pe | |
| 1,2-difluoro-4-methoxybenzene |
Safety and Handling
TSCA : No