accessibility menu, dialog, popup

UserName

CAS RN 115144-40-6

IUPAC Name: 1,2-difluoro-4-methoxybenzene
Synonyms: 1,2-difluoro-4-methoxybenzene
Molecular Weight (g/mol): 144.12
Molecular Formula: C7H6F2O
InChi Key: DSEVNUCNUTYYHW-UHFFFAOYSA-N
SMILES: COC1=CC=C(F)C(F)=C1
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)