missing translation for 'onlineSavingsMsg'
Learn More

3,4-Difluoroanisole 97.0+%, TCI America™

Catalog No. D28565G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D28565G 5 g
D285625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D28565G Supplier TCI America Supplier No. D28565G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 115144-40-6
Molecular Formula C7H6F2O
Molecular Weight (g/mol) 144.121
MDL Number MFCD00010730
InChI Key DSEVNUCNUTYYHW-UHFFFAOYSA-N
Synonym 3,4-difluoroanisole, 1,2-difluoro-4-methoxy-benzene, benzene, 1,2-difluoro-4-methoxy, 3,4-difluoro-1-methoxybenzene, 1,2-difluor-4-methoxybenzol, pubchem3457, 3,4-difluoro-anisole, difluorobenzene,1-methoxy, acmc-2099pe
PubChem CID 568250
IUPAC Name 1,2-difluoro-4-methoxybenzene
SMILES COC1=CC(=C(C=C1)F)F

Specifications

Color Yellow
Quantity 5 g
UN Number 1993
Formula Weight 144.12
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 3,4-Difluoroanisole