missing translation for 'onlineSavingsMsg'
Learn More

3,4-Difluoroanisole, 98%

Catalog No. aaa1247414
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
100 g
25 g
5 g
Catalog No. Quantity
AAA1247414 25 g
AAA1247406 5 g
AAA1247422 100 g
3 options

Catalog No. AAA1247414

Supplier: Thermo Scientific Chemicals A1247414

Only null left

CAS: 115144-40-6 | C7H6F2O | 144.121 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 115144-40-6
Molecular Formula C7H6F2O
Molecular Weight (g/mol) 144.121
MDL Number MFCD00010730
InChI Key DSEVNUCNUTYYHW-UHFFFAOYSA-N
Synonym 3,4-difluoroanisole, 1,2-difluoro-4-methoxy-benzene, benzene, 1,2-difluoro-4-methoxy, 3,4-difluoro-1-methoxybenzene, 1,2-difluor-4-methoxybenzol, pubchem3457, 3,4-difluoro-anisole, difluorobenzene,1-methoxy, acmc-2099pe
PubChem CID 568250
IUPAC Name 1,2-difluoro-4-methoxybenzene
SMILES COC1=CC(=C(C=C1)F)F

Specifications

Density 1.214
Flash Point 55°C (131°F)
Refractive Index 1.468
Quantity 25 g
UN Number UN1993
Beilstein 6717610
Formula Weight 144.12
Percent Purity 98%
Chemical Name or Material 3,4-Difluoroanisole
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only