missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dichlorophenylacetonitrile, 98%

Catalog No. AAA1138214
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1138214 25 g
AAA1138222 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1138214 Supplier Thermo Scientific Chemicals Supplier No. A1138214
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 3218-49-3 | C8H5Cl2N | 186.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3218-49-3
Molecular Formula C8H5Cl2N
Molecular Weight (g/mol) 186.04
MDL Number MFCD00001909
InChI Key QWZNCAFWRZZJMA-UHFFFAOYSA-N
Synonym 3,4-dichlorophenylacetonitrile, 2-3,4-dichlorophenyl acetonitrile, 3,4-dichlorobenzyl cyanide, 3,4-dichlorophenyl acetonitrile, benzeneacetonitrile, 3,4-dichloro, 3,4-dichlorobenzeneacetonitrile, 2-3,4-dichlorophenyl ethanenitrile, 3,4-dichlorphenyl acetonitrile, pubchem16506, 3,4-dichlorobenzylcyanide
PubChem CID 76690
IUPAC Name 2-(3,4-dichlorophenyl)acetonitrile
SMILES ClC1=CC=C(CC#N)C=C1Cl

Specifications

Melting Point 38°C to 42°C
Boiling Point 169°C to 171°C (12 mmHg)
Flash Point >110°C (230°F)
Odor Characteristic
Quantity 25 g
UN Number UN3439
Beilstein 638749
Formula Weight 186.04
Percent Purity 98%
Chemical Name or Material 3,4-Dichlorophenylacetonitrile
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H301-H311-H331-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
EINECSNumber 221-743-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only