accessibility menu, dialog, popup

UserName

CAS RN 3218-49-3

ClNCl

CAS RN 3218-49-3

IUPAC Name: 2-(3,4-dichlorophenyl)acetonitrile
Synonyms: 2-(3,4-dichlorophenyl)acetonitrile
Molecular Weight (g/mol): 186.04
Molecular Formula: C8H5Cl2N
InChi Key: QWZNCAFWRZZJMA-UHFFFAOYSA-N
SMILES: ClC1=CC=C(CC#N)C=C1Cl
Boiling Point 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)