missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dichlorobenzyl Cyanide 98.0+%, TCI America™

Catalog No. D179425G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D179425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D179425G Supplier TCI America Supplier No. D179425G

Chemical Identifiers

CAS 3218-49-3
Molecular Formula C8H5Cl2N
Molecular Weight (g/mol) 186.04
MDL Number MFCD00001909
InChI Key QWZNCAFWRZZJMA-UHFFFAOYSA-N
Synonym 3,4-dichlorophenylacetonitrile, 2-3,4-dichlorophenyl acetonitrile, 3,4-dichlorobenzyl cyanide, 3,4-dichlorophenyl acetonitrile, benzeneacetonitrile, 3,4-dichloro, 3,4-dichlorobenzeneacetonitrile, 2-3,4-dichlorophenyl ethanenitrile, 3,4-dichlorphenyl acetonitrile, pubchem16506, 3,4-dichlorobenzylcyanide
PubChem CID 76690
IUPAC Name 2-(3,4-dichlorophenyl)acetonitrile
SMILES ClC1=CC=C(CC#N)C=C1Cl

Specifications

Melting Point 40°C
Color White
Boiling Point 171°C
Quantity 25 g
UN Number 3439
Formula Weight 186.04
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,4-Dichlorobenzyl Cyanide
EINECSNumber (3)-3822
TSCA No