missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dichloro-6-fluoroaniline, 96%

Catalog No. p-7039967
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH59506MD 250 mg
AAH5950603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH59506MD Supplier Thermo Scientific Chemicals Supplier No. H59506MD
Only null left

CAS: 2729-36-4 | C6H4Cl2FN | 180.003 g/mol

3,4-Dichloro-6-fluoroaniline is used as pharmaceutical intermediates

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,4-Dichloro-6-fluoroaniline is used as pharmaceutical intermediates

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 2729-36-4
Molecular Formula C6H4Cl2FN
Molecular Weight (g/mol) 180.003
MDL Number MFCD19707630
InChI Key FHJVHVKVTUTGTA-UHFFFAOYSA-N
Synonym 3,4-dichloro-6-fluoroaniline, 4,5-dichloro-2-fluorobenzenamine, 4,5-dichloro-2-fluorophenylamine, 4,5-dichloro-2-fluoro-phenylamine, 4,5-dichloro-2-fluorophenyl amine, benzenamine, 4,5-dichloro-2-fluoro
PubChem CID 223091
IUPAC Name 4,5-dichloro-2-fluoroaniline
SMILES C1=C(C(=CC(=C1Cl)Cl)F)N

Specifications

Quantity 250 mg
UN Number UN2811
Formula Weight 180.01
Percent Purity 96%
Chemical Name or Material 3,4-Dichloro-6-fluoroaniline
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only