missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,4-Dibutoxy-3-cyclobutene-1,2-dione 97.0+%, TCI America™
Supplier: TCI America D22035G
Specifications
| 3,4-Dibutoxy-3-cyclobutene-1,2-dione | |
| Yellow | |
| MFCD00037150 | |
| dibutyl squarate, 3,4-dibutoxy-3-cyclobutene-1,2-dione, squaric acid dibutyl ester, sadbe, squaric acid dibutylester, 3,4-di-n-butoxy-3-cyclobutene-1,2-dione, unii-4rto57vg65, 1,2-dibutyl squarate, 3-cyclobutene-1,2-dione, 3,4-dibutoxy, acmc-209h5p | |
| CCCCOC1=C(OCCCC)C(=O)C1=O | |
| 226.27 | |
| CHEBI:53612 | |
| ≥97.0% (GC) |
| 2892-62-8 | |
| C12H18O4 | |
| 5 g | |
| XBRWELTXMQSEIN-UHFFFAOYSA-N | |
| dibutoxycyclobut-3-ene-1,2-dione | |
| 65108 | |
| 226.27 | |
| Liquid |
Chemical Identifiers
| 2892-62-8 | |
| 226.27 | |
| XBRWELTXMQSEIN-UHFFFAOYSA-N | |
| 65108 | |
| dibutoxycyclobut-3-ene-1,2-dione |
| C12H18O4 | |
| MFCD00037150 | |
| dibutyl squarate, 3,4-dibutoxy-3-cyclobutene-1,2-dione, squaric acid dibutyl ester, sadbe, squaric acid dibutylester, 3,4-di-n-butoxy-3-cyclobutene-1,2-dione, unii-4rto57vg65, 1,2-dibutyl squarate, 3-cyclobutene-1,2-dione, 3,4-dibutoxy, acmc-209h5p | |
| CHEBI:53612 | |
| CCCCOC1=C(OCCCC)C(=O)C1=O |
Safety and Handling
RTECSNumber : GU1772066
TSCA : No