missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dibutoxy-3-cyclobutene-1,2-dione 97.0+%, TCI America™

Catalog No. p-7127167
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D22035G 5 g
D220325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D22035G Supplier TCI America Supplier No. D22035G

Chemical Identifiers

CAS 2892-62-8
Molecular Formula C12H18O4
Molecular Weight (g/mol) 226.27
MDL Number MFCD00037150
InChI Key XBRWELTXMQSEIN-UHFFFAOYSA-N
Synonym dibutyl squarate, 3,4-dibutoxy-3-cyclobutene-1,2-dione, squaric acid dibutyl ester, sadbe, squaric acid dibutylester, 3,4-di-n-butoxy-3-cyclobutene-1,2-dione, unii-4rto57vg65, 1,2-dibutyl squarate, 3-cyclobutene-1,2-dione, 3,4-dibutoxy, acmc-209h5p
PubChem CID 65108
ChEBI CHEBI:53612
IUPAC Name dibutoxycyclobut-3-ene-1,2-dione
SMILES CCCCOC1=C(OCCCC)C(=O)C1=O

Specifications

Color Yellow
Quantity 5 g
Formula Weight 226.27
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 3,4-Dibutoxy-3-cyclobutene-1,2-dione
RTECSNumber GU1772066
TSCA No