missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dibromosulfolane 98.0+%, TCI America™

Catalog No. D067925G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D067925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D067925G Supplier TCI America Supplier No. D067925G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 15091-30-2
Molecular Formula C4H6Br2O2S
Molecular Weight (g/mol) 277.958
MDL Number MFCD00022536
InChI Key CGWKQCYAMPDEGC-UHFFFAOYSA-N
Synonym 3,4-Dibromotetrahydrothiophene 1,1-Dioxide
PubChem CID 85797
IUPAC Name 3,4-dibromothiolane 1,1-dioxide
SMILES C1C(C(CS1(=O)=O)Br)Br

Specifications

Melting Point 143°C
Color White
Quantity 25 g
Formula Weight 277.96
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,4-Dibromosulfolane
RTECSNumber XN0470000
TSCA Yes