accessibility menu, dialog, popup

UserName

BrSOOBr

CAS RN 15091-30-2

IUPAC Name: 3,4-dibromo-1λ⁶-thiolane-1,1-dione
Synonyms: 3,4-dibromo-1λ⁶-thiolane-1,1-dione
Molecular Weight (g/mol): 277.96
Molecular Formula: C4H6Br2O2S
InChi Key: CGWKQCYAMPDEGC-UHFFFAOYNA-N
SMILES: BrC1CS(=O)(=O)CC1Br
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results