missing translation for 'onlineSavingsMsg'
Learn More

3-(4-Carboxyphenyl)propionic acid, 98%

CAS: 38628-51-2 | C10H10O4 | 194.19 g/mol

$187.12 - $2,128.96

Chemical Identifiers

CAS 38628-51-2
Molecular Formula C10H10O4
Molecular Weight (g/mol) 194.19
MDL Number MFCD00016560
InChI Key QSPCOYVUYYFWAU-UHFFFAOYSA-N
Synonym 4-2-carboxyethyl benzoic acid, 3-4-carboxyphenyl propionic acid, 3-4-carboxyphenyl propanoic acid, benzenepropanoic acid, 4-carboxy, pubchem19268, acmc-1ctf7, 4-carboxyhydrocinnamic acid, 4-carboxybenzenepropionic acid, 4-2-carboxyethyl benzoicacid, carboxyphenylpropionicacid; 985
PubChem CID 854054
IUPAC Name 4-(2-carboxyethyl)benzoic acid
SMILES OC(=O)CCC1=CC=C(C=C1)C(O)=O
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1508106
View Documents
Thermo Scientific Chemicals
A1508106
5 g N/A
Only null left
 
AAA1508109
View Documents
Thermo Scientific Chemicals
A1508109
10 g N/A
Only null left
 
AAA1508114
View Documents
Thermo Scientific Chemicals
A1508114
25 g N/A
Only null left
 
AAA1508122
View Documents
Thermo Scientific Chemicals
A1508122
100 g N/A
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

38628-51-2
194.19
QSPCOYVUYYFWAU-UHFFFAOYSA-N
854054
OC(=O)CCC1=CC=C(C=C1)C(O)=O
C10H10O4
MFCD00016560
4-2-carboxyethyl benzoic acid, 3-4-carboxyphenyl propionic acid, 3-4-carboxyphenyl propanoic acid, benzenepropanoic acid, 4-carboxy, pubchem19268, acmc-1ctf7, 4-carboxyhydrocinnamic acid, 4-carboxybenzenepropionic acid, 4-2-carboxyethyl benzoicacid, carboxyphenylpropionicacid; 985
4-(2-carboxyethyl)benzoic acid

Specifications

38628-51-2
C10H10O4
5 g
4-2-carboxyethyl benzoic acid, 3-4-carboxyphenyl propionic acid, 3-4-carboxyphenyl propanoic acid, benzenepropanoic acid, 4-carboxy, pubchem19268, acmc-1ctf7, 4-carboxyhydrocinnamic acid, 4-carboxybenzenepropionic acid, 4-2-carboxyethyl benzoicacid, carboxyphenylpropionicacid; 985
OC(=O)CCC1=CC=C(C=C1)C(O)=O
194.19
194.19
3-(4-Carboxyphenyl)propionic acid
288°C to 291°C
MFCD00016560
3271796
QSPCOYVUYYFWAU-UHFFFAOYSA-N
4-(2-carboxyethyl)benzoic acid
854054
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only