missing translation for 'onlineSavingsMsg'
Learn More

3-(4-Carboxyphenyl)propionic acid, 98%

CAS: 38628-51-2 | C10H10O4 | 194.19 g/mol

Supplier:  Thermo Scientific Chemicals A1508109

 View more versions of this product

Catalog No. AAA1508109


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

3-(4-Carboxyphenyl)propionic acid
288°C to 291°C
MFCD00016560
3271796
QSPCOYVUYYFWAU-UHFFFAOYSA-N
4-(2-carboxyethyl)benzoic acid
854054
98%
38628-51-2
C10H10O4
10 g
4-2-carboxyethyl benzoic acid, 3-4-carboxyphenyl propionic acid, 3-4-carboxyphenyl propanoic acid, benzenepropanoic acid, 4-carboxy, pubchem19268, acmc-1ctf7, 4-carboxyhydrocinnamic acid, 4-carboxybenzenepropionic acid, 4-2-carboxyethyl benzoicacid, carboxyphenylpropionicacid; 985
OC(=O)CCC1=CC=C(C=C1)C(O)=O
194.19
194.19

Chemical Identifiers

38628-51-2
194.19
QSPCOYVUYYFWAU-UHFFFAOYSA-N
854054
OC(=O)CCC1=CC=C(C=C1)C(O)=O
C10H10O4
MFCD00016560
4-2-carboxyethyl benzoic acid, 3-4-carboxyphenyl propionic acid, 3-4-carboxyphenyl propanoic acid, benzenepropanoic acid, 4-carboxy, pubchem19268, acmc-1ctf7, 4-carboxyhydrocinnamic acid, 4-carboxybenzenepropionic acid, 4-2-carboxyethyl benzoicacid, carboxyphenylpropionicacid; 985
4-(2-carboxyethyl)benzoic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only