accessibility menu, dialog, popup

UserName

OHOOHO

CAS RN 38628-51-2

IUPAC Name: 4-(2-carboxyethyl)benzoic acid
Synonyms: 4-(2-carboxyethyl)benzoic acid
Molecular Weight (g/mol): 194.19
Molecular Formula: C10H10O4
InChi Key: QSPCOYVUYYFWAU-UHFFFAOYSA-N
SMILES: OC(=O)CCC1=CC=C(C=C1)C(O)=O
Percent Purity 
  • (1)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results