missing translation for 'onlineSavingsMsg'
Learn More

3,4,6-Tri-O-benzyl-D-galactal, Thermo Scientific Chemicals

$631.97 - $631.97

Chemical Identifiers

CAS 80040-79-5
Molecular Formula C27H28O4
Molecular Weight (g/mol) 416.52
MDL Number MFCD09039276
InChI Key MXYLLYBWXIUMIT-ZONZVBGPSA-N
Synonym tri-o-benzyl-d-galactal, 3,4,6-tri-o-benzyl-d-galactal, 2r,3r,4r-3,4-bis benzyloxy-2-benzyloxy methyl-3,4-dihydro-2h-pyran, 3-o,4-o,6-o-tribenzyl-1,2-dideoxy-d-lyxo-1-hexenopyranose, 1,5-anhydro-2-deoxy-d-lyxo-hex-1-enitol 3,4,6-tribenzyl ether, 2r,3r,4r-3,4-bis phenylmethoxy-2-phenylmethoxymethyl-3,4-dihydro-2h-pyran
PubChem CID 2734741
IUPAC Name (2R,3R,4R)-3,4-bis(benzyloxy)-2-[(benzyloxy)methyl]-3,4-dihydro-2H-pyran
SMILES C(OCC1=CC=CC=C1)[C@H]1OC=C[C@@H](OCC2=CC=CC=C2)[C@H]1OCC1=CC=CC=C1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ6620206
View Documents
Thermo Scientific Chemicals
J6620206
5 g
Each for $631.97
Only null left
 
Description

Description

An important building block for solution- and solid-phase synthesis of oligosaccharides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

80040-79-5
416.52
MXYLLYBWXIUMIT-ZONZVBGPSA-N
2734741
C(OCC1=CC=CC=C1)[C@H]1OC=C[C@@H](OCC2=CC=CC=C2)[C@H]1OCC1=CC=CC=C1
C27H28O4
MFCD09039276
tri-o-benzyl-d-galactal, 3,4,6-tri-o-benzyl-d-galactal, 2r,3r,4r-3,4-bis benzyloxy-2-benzyloxy methyl-3,4-dihydro-2h-pyran, 3-o,4-o,6-o-tribenzyl-1,2-dideoxy-d-lyxo-1-hexenopyranose, 1,5-anhydro-2-deoxy-d-lyxo-hex-1-enitol 3,4,6-tribenzyl ether, 2r,3r,4r-3,4-bis phenylmethoxy-2-phenylmethoxymethyl-3,4-dihydro-2h-pyran
(2R,3R,4R)-3,4-bis(benzyloxy)-2-[(benzyloxy)methyl]-3,4-dihydro-2H-pyran

Specifications

80040-79-5
White to Yellow
C27H28O4
5 g
Soluble in dichloromethane or chloroform,
−43° (c=1 in Dichloromethane)
(2R,3R,4R)-3,4-bis(benzyloxy)-2-[(benzyloxy)methyl]-3,4-dihydro-2H-pyran
2734741
Powder
48°C to 54°C
>110°C (235°F)
MFCD09039276
tri-o-benzyl-d-galactal, 3,4,6-tri-o-benzyl-d-galactal, 2r,3r,4r-3,4-bis benzyloxy-2-benzyloxy methyl-3,4-dihydro-2h-pyran, 3-o,4-o,6-o-tribenzyl-1,2-dideoxy-d-lyxo-1-hexenopyranose, 1,5-anhydro-2-deoxy-d-lyxo-hex-1-enitol 3,4,6-tribenzyl ether, 2r,3r,4r-3,4-bis phenylmethoxy-2-phenylmethoxymethyl-3,4-dihydro-2h-pyran
MXYLLYBWXIUMIT-ZONZVBGPSA-N
C(OCC1=CC=CC=C1)[C@H]1OC=C[C@@H](OCC2=CC=CC=C2)[C@H]1OCC1=CC=CC=C1
416.52
416.51
3,4,6-Tri-O-benzyl-D-galactal
Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Store at -20°C

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only