missing translation for 'onlineSavingsMsg'
Learn More

3,4,5-Trimethoxyphenylacetonitrile 97.0+%, TCI America™

Catalog No. p-7137025
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T19275G 5 g
T192725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T19275G Supplier TCI America Supplier No. T19275G
Only null left

Chemical Identifiers

CAS 13338-63-1
Molecular Formula C11H13NO3
Molecular Weight (g/mol) 207.23
MDL Number MFCD00001912
InChI Key ACFJNTXCEQCDBX-UHFFFAOYSA-N
Synonym 3,4,5-trimethoxyphenylacetonitrile, 2-3,4,5-trimethoxyphenyl acetonitrile, 3,4,5-trimethoxybenzyl cyanide, 3,4,5-trimethoxybenzylnitrile, 3,4,5-trimethoxybenzeneacetonitrile, benzeneacetonitrile, 3,4,5-trimethoxy, 3,4,5-trimethoxyphenylacetylnitryl, 3,4,5-trimethoxyphenyl acetonitrile, acetonitrile, 3,4,5-trimethoxyphenyl, 2-3,4,5-trimethoxyphenyl ethanenitrile
PubChem CID 25887
IUPAC Name 2-(3,4,5-trimethoxyphenyl)acetonitrile
SMILES COC1=CC(CC#N)=CC(OC)=C1OC

Specifications

Color White
Quantity 5 g
UN Number 3439
Formula Weight 207.23
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,4,5-Trimethoxyphenylacetonitrile
RTECSNumber AM2475000
TSCA No