accessibility menu, dialog, popup

UserName

CAS RN 13338-63-1

IUPAC Name: 2-(3,4,5-trimethoxyphenyl)acetonitrile
Synonyms: 2-(3,4,5-trimethoxyphenyl)acetonitrile
Molecular Weight (g/mol): 207.23
Molecular Formula: C11H13NO3
InChi Key: ACFJNTXCEQCDBX-UHFFFAOYSA-N
SMILES: COC1=CC(CC#N)=CC(OC)=C1OC
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)