missing translation for 'onlineSavingsMsg'
Learn More

3,4,5-Trimethoxybenzylamine, 96%

CAS: 18638-99-8 | C10H15NO3 | 197.234 g/mol

Supplier:  Thermo Scientific Chemicals L1346106

 View more versions of this product

Catalog No. AAL1346106


Only null left
Explore available promotions

Description

Description

3,4,5-Trimethoxybenzylamine is used in the synthesis of benzyl 3-deoxy-3-(3,4,5-trimethoxybenzylamino)-β-L-xylopyranoside. It is a important organic intermediate. It is used in agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,4,5-Trimethoxybenzylamine is used in the synthesis of benzyl 3-deoxy-3-(3,4,5-trimethoxybenzylamino)-β-L-xylopyranoside. It is a important organic intermediate. It is used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Not miscible or difficult to mix in water.

Notes
It is sensitive to air. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
Specifications

Specifications

3,4,5-Trimethoxybenzylamine
1.155
>110°C (230°F)
1.548
5 g
1107570
3,4,5-trimethoxybenzylamine, 3,4,5-trimethoxyphenyl methanamine, 1-3,4,5-trimethoxyphenyl methanamine, benzenemethanamine, 3,4,5-trimethoxy, 3,4,5-trimethoxyphenyl methylamine, 1-3,4,5-trimethoxyphenyl methanamine hydrochloride, pubchem16674, acmc-1c5ez, ksc495e6b, 3,4,5-trimethoxy benzylamine
YUPUSBMJCFBHAP-UHFFFAOYSA-N
(3,4,5-trimethoxyphenyl)methanamine
87736
96%
18638-99-8
120°C to 122°C (0.05 mmHg)
C10H15NO3
MFCD00017150
UN2735
Air Sensitive
Not miscible or difficult to mix in water.
COC1=CC(=CC(=C1OC)OC)CN
197.234
197.24

Chemical Identifiers

18638-99-8
197.234
YUPUSBMJCFBHAP-UHFFFAOYSA-N
87736
COC1=CC(=CC(=C1OC)OC)CN
C10H15NO3
MFCD00017150
3,4,5-trimethoxybenzylamine, 3,4,5-trimethoxyphenyl methanamine, 1-3,4,5-trimethoxyphenyl methanamine, benzenemethanamine, 3,4,5-trimethoxy, 3,4,5-trimethoxyphenyl methylamine, 1-3,4,5-trimethoxyphenyl methanamine hydrochloride, pubchem16674, acmc-1c5ez, ksc495e6b, 3,4,5-trimethoxy benzylamine
(3,4,5-trimethoxyphenyl)methanamine
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.

EINECSNumber : 242-468-0

TSCA : No

Recommended Storage : Ambient temperatures; Store under Argon

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only