missing translation for 'onlineSavingsMsg'
Learn More

3,4,5-Trimethoxybenzylamine, 96%

Catalog No. p-7046208
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1346106 5 g
AAL1346114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1346106 Supplier Thermo Scientific Chemicals Supplier No. L1346106
Only null left

CAS: 18638-99-8 | C10H15NO3 | 197.234 g/mol

3,4,5-Trimethoxybenzylamine is used in the synthesis of benzyl 3-deoxy-3-(3,4,5-trimethoxybenzylamino)-β-L-xylopyranoside. It is a important organic intermediate. It is used in agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,4,5-Trimethoxybenzylamine is used in the synthesis of benzyl 3-deoxy-3-(3,4,5-trimethoxybenzylamino)-β-L-xylopyranoside. It is a important organic intermediate. It is used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Not miscible or difficult to mix in water.

Notes
It is sensitive to air. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 18638-99-8
Molecular Formula C10H15NO3
Molecular Weight (g/mol) 197.234
MDL Number MFCD00017150
InChI Key YUPUSBMJCFBHAP-UHFFFAOYSA-N
Synonym 3,4,5-trimethoxybenzylamine, 3,4,5-trimethoxyphenyl methanamine, 1-3,4,5-trimethoxyphenyl methanamine, benzenemethanamine, 3,4,5-trimethoxy, 3,4,5-trimethoxyphenyl methylamine, 1-3,4,5-trimethoxyphenyl methanamine hydrochloride, pubchem16674, acmc-1c5ez, ksc495e6b, 3,4,5-trimethoxy benzylamine
PubChem CID 87736
IUPAC Name (3,4,5-trimethoxyphenyl)methanamine
SMILES COC1=CC(=CC(=C1OC)OC)CN

Specifications

Density 1.155
Boiling Point 120°C to 122°C (0.05 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.548
Quantity 5 g
UN Number UN2735
Beilstein 1107570
Sensitivity Air Sensitive
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 197.24
Percent Purity 96%
Chemical Name or Material 3,4,5-Trimethoxybenzylamine
Show More Show Less
Hazard Category H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
EINECSNumber 242-468-0
TSCA No
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only