missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,4,5-Trimethoxybenzyl Alcohol 98.0+%, TCI America™
$65.24 - $1,038.29
Chemical Identifiers
| CAS | 3840-31-1 |
|---|---|
| Molecular Formula | C10H14O4 |
| Molecular Weight (g/mol) | 198.218 |
| MDL Number | MFCD00004639 |
| InChI Key | QPHLRCUCFDXGLY-UHFFFAOYSA-N |
| Synonym | 3,4,5-trimethoxybenzyl alcohol, 3,4,5-trimethoxyphenyl methanol, benzenemethanol, 3,4,5-trimethoxy, 3,4,5-trimethoxybenzylalcohol, 3,4,5-trimethoxybenzylic alcohol, benzyl alcohol, 3,4,5-trimethoxy, 3,4,5-trimethoxyphenyl methan-1-ol, acmc-1cp7y, 3,4,5-trimethoxybenzalcohol, ksc494s0n |
| PubChem CID | 77449 |
| IUPAC Name | (3,4,5-trimethoxyphenyl)methanol |
| SMILES | COC1=CC(=CC(=C1OC)OC)CO |
Chemical Identifiers
| 3840-31-1 | |
| 198.218 | |
| QPHLRCUCFDXGLY-UHFFFAOYSA-N | |
| 77449 | |
| COC1=CC(=CC(=C1OC)OC)CO |
| C10H14O4 | |
| MFCD00004639 | |
| 3,4,5-trimethoxybenzyl alcohol, 3,4,5-trimethoxyphenyl methanol, benzenemethanol, 3,4,5-trimethoxy, 3,4,5-trimethoxybenzylalcohol, 3,4,5-trimethoxybenzylic alcohol, benzyl alcohol, 3,4,5-trimethoxy, 3,4,5-trimethoxyphenyl methan-1-ol, acmc-1cp7y, 3,4,5-trimethoxybenzalcohol, ksc494s0n | |
| (3,4,5-trimethoxyphenyl)methanol |
Specifications
| 3840-31-1 | |
| 155°C | |
| MFCD00004639 | |
| 3,4,5-trimethoxybenzyl alcohol, 3,4,5-trimethoxyphenyl methanol, benzenemethanol, 3,4,5-trimethoxy, 3,4,5-trimethoxybenzylalcohol, 3,4,5-trimethoxybenzylic alcohol, benzyl alcohol, 3,4,5-trimethoxy, 3,4,5-trimethoxyphenyl methan-1-ol, acmc-1cp7y, 3,4,5-trimethoxybenzalcohol, ksc494s0n | |
| COC1=CC(=CC(=C1OC)OC)CO | |
| 198.218 | |
| 198.22 | |
| Liquid |
| Yellow | |
| C10H14O4 | |
| 25 g | |
| QPHLRCUCFDXGLY-UHFFFAOYSA-N | |
| (3,4,5-trimethoxyphenyl)methanol | |
| 77449 | |
| ≥98.0% (GC) | |
| 3,4,5-Trimethoxybenzyl Alcohol |
Safety and Handling
TSCA : No