accessibility menu, dialog, popup

UserName

H3COOHOH3COH3C

CAS RN 3840-31-1

IUPAC Name: (3,4,5-trimethoxyphenyl)methanol
Synonyms: (3,4,5-trimethoxyphenyl)methanol
Molecular Weight (g/mol): 198.22
Molecular Formula: C10H14O4
InChi Key: QPHLRCUCFDXGLY-UHFFFAOYSA-N
SMILES: COC1=CC(CO)=CC(OC)=C1OC
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)