missing translation for 'onlineSavingsMsg'
Learn More

3,4,5-Trimethoxybenzyl Alcohol 98.0+%, TCI America™

Catalog No. T110325G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T110325G 25 g
T1103500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T110325G Supplier TCI America Supplier No. T110325G

Chemical Identifiers

CAS 3840-31-1
Molecular Formula C10H14O4
Molecular Weight (g/mol) 198.218
MDL Number MFCD00004639
InChI Key QPHLRCUCFDXGLY-UHFFFAOYSA-N
Synonym 3,4,5-trimethoxybenzyl alcohol, 3,4,5-trimethoxyphenyl methanol, benzenemethanol, 3,4,5-trimethoxy, 3,4,5-trimethoxybenzylalcohol, 3,4,5-trimethoxybenzylic alcohol, benzyl alcohol, 3,4,5-trimethoxy, 3,4,5-trimethoxyphenyl methan-1-ol, acmc-1cp7y, 3,4,5-trimethoxybenzalcohol, ksc494s0n
PubChem CID 77449
IUPAC Name (3,4,5-trimethoxyphenyl)methanol
SMILES COC1=CC(=CC(=C1OC)OC)CO

Specifications

Color Yellow
Boiling Point 155°C
Quantity 25 g
Formula Weight 198.22
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3,4,5-Trimethoxybenzyl Alcohol