missing translation for 'onlineSavingsMsg'
Learn More

3,4,5-Trimethoxybenzyl alcohol, 98%

Catalog No. p-7020945
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1307109 10 g
AAA1307118 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1307109 Supplier Thermo Scientific Chemicals Supplier No. A1307109

CAS: 3840-31-1 | C10H14O4 | 198.218 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3840-31-1
Molecular Formula C10H14O4
Molecular Weight (g/mol) 198.218
MDL Number MFCD00004639
InChI Key QPHLRCUCFDXGLY-UHFFFAOYSA-N
Synonym 3,4,5-trimethoxybenzyl alcohol, 3,4,5-trimethoxyphenyl methanol, benzenemethanol, 3,4,5-trimethoxy, 3,4,5-trimethoxybenzylalcohol, 3,4,5-trimethoxybenzylic alcohol, benzyl alcohol, 3,4,5-trimethoxy, 3,4,5-trimethoxyphenyl methan-1-ol, acmc-1cp7y, 3,4,5-trimethoxybenzalcohol, ksc494s0n
PubChem CID 77449
IUPAC Name (3,4,5-trimethoxyphenyl)methanol
SMILES COC1=CC(=CC(=C1OC)OC)CO

Specifications

Density 1.199
Boiling Point 228°C (25 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.5445
Quantity 10 g
Beilstein 2104523
Formula Weight 198.22
Percent Purity 98%
Chemical Name or Material 3,4,5-Trimethoxybenzyl alcohol
EINECSNumber 223-331-4
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only