missing translation for 'onlineSavingsMsg'
Learn More

3,4,5-Trifluoronitrobenzene 98.0+%, TCI America™

Catalog No. p-7137177
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T23575G 5 g
T235725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T23575G Supplier TCI America Supplier No. T23575G
Only null left

Chemical Identifiers

CAS 66684-58-0
Molecular Formula C6H2F3NO2
Molecular Weight (g/mol) 177.08
MDL Number MFCD00456803
InChI Key PTTUMBGORBMNBN-UHFFFAOYSA-N
Synonym 3,4,5-trifluoronitrobenzene, 1-nitro-3,4,5-trifluorobenzene, 3,4,5-trifluoro-nitrobenzene, 1,2,3-trifluoro-5-nitro-benzene, benzene, 1,2,3-trifluoro-5-nitro, pubchem1071, acmc-1baej, 3,4,5-trifluronitrobenzene, ksc495s3n, 3,4,5-trifluoronitro benzene
PubChem CID 2782793
IUPAC Name 1,2,3-trifluoro-5-nitrobenzene
SMILES [O-][N+](=O)C1=CC(F)=C(F)C(F)=C1

Specifications

Color Yellow
Quantity 5 g
Formula Weight 177.08
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3,4,5-Trifluoronitrobenzene