missing translation for 'onlineSavingsMsg'
Learn More

3,4,5-Trifluorobenzyl Bromide 97.0+%, TCI America™

Catalog No. p-7137228
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T24641G 1 g
T24645G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T24641G Supplier TCI America Supplier No. T24641G
Only null left

Chemical Identifiers

CAS 220141-72-0
Molecular Formula C7H4BrF3
Molecular Weight (g/mol) 225.01
MDL Number MFCD00083528
InChI Key MPEKZIYOLQCWLL-UHFFFAOYSA-N
Synonym 3,4,5-trifluorobenzyl bromide, 5-bromomethyl-1,2,3-trifluorobenzene, 3,4,5-trifluorobenzylbromide, alpha-bromo-3,4,5-trifluorotoluene, 3,4,5-trifluoro benzyl bromide, benzene, 5-bromomethyl-1,2,3-trifluoro, a-bromo-3,4,5-trifluorotoluene, pubchem4670, acmc-1cupe
PubChem CID 2777062
IUPAC Name 5-(bromomethyl)-1,2,3-trifluorobenzene
SMILES FC1=CC(CBr)=CC(F)=C1F

Specifications

Boiling Point 46°C
Quantity 1 g
UN Number 3265
Formula Weight 225.01
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 3,4,5-Trifluorobenzyl Bromide