accessibility menu, dialog, popup

UserName

FBrFF

CAS RN 220141-72-0

IUPAC Name: 5-(bromomethyl)-1,2,3-trifluorobenzene
Synonyms: 5-(bromomethyl)-1,2,3-trifluorobenzene
Molecular Weight (g/mol): 225.01
Molecular Formula: C7H4BrF3
InChi Key: MPEKZIYOLQCWLL-UHFFFAOYSA-N
SMILES: FC1=CC(CBr)=CC(F)=C1F
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)