missing translation for 'onlineSavingsMsg'
Learn More

3,4,5-Trifluorobenzyl bromide, 97%

Catalog No. AAB2257803
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2257803 1 g
AAB2257806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2257803 Supplier Thermo Scientific Chemicals Supplier No. B2257803

CAS: 220141-72-0 | C7H4BrF3 | 225.01 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 220141-72-0
Molecular Formula C7H4BrF3
Molecular Weight (g/mol) 225.01
MDL Number MFCD00083528
InChI Key MPEKZIYOLQCWLL-UHFFFAOYSA-N
Synonym 3,4,5-trifluorobenzyl bromide, 5-bromomethyl-1,2,3-trifluorobenzene, 3,4,5-trifluorobenzylbromide, alpha-bromo-3,4,5-trifluorotoluene, 3,4,5-trifluoro benzyl bromide, benzene, 5-bromomethyl-1,2,3-trifluoro, a-bromo-3,4,5-trifluorotoluene, pubchem4670, acmc-1cupe
PubChem CID 2777062
IUPAC Name 5-(bromomethyl)-1,2,3-trifluorobenzene
SMILES FC1=CC(CBr)=CC(F)=C1F

Specifications

Density 1.689
Boiling Point 44°C to 46°C (1 mmHg)
Flash Point 65°C (149°F)
Refractive Index 1.505
Quantity 1 g
UN Number UN3265
Beilstein 8680985
Formula Weight 225.01
Percent Purity 97%
Chemical Name or Material 3,4,5-Trifluorobenzyl bromide
Hazard Category H227-H314-H335
Hazard Statement GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Precautionary Statement P210-P235-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only