Learn More
3,4,5-Trifluorobenzeneboronic acid, 97%
CAS: 143418-49-9 | C6H4BF3O2 | 175.901 g/mol
Supplier: Thermo Scientific Chemicals L18519MD
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 3,4,5-Trifluorobenzeneboronic acid | |
| 275°C to 279°C | |
| MFCD02093069 | |
| 7371914 | |
| UHDDEIOYXFXNNJ-UHFFFAOYSA-N | |
| (3,4,5-trifluorophenyl)boronic acid | |
| 2734671 | |
| 97% |
| 143418-49-9 | |
| C6H4BF3O2 | |
| 250 mg | |
| 3,4,5-trifluorophenyl boronic acid, 3,4,5-trifluorobenzeneboronic acid, 3,4,5-trifluorophenyl boranediol, boronic acid, b-3,4,5-trifluorophenyl, 3,4,5-trifluoro phenylboric acid, boronic acid, 3,4,5-trifluorophenyl, 3,4,5-trifluorophenylboronicacid, pubchem2340, acmc-209cqj | |
| B(C1=CC(=C(C(=C1)F)F)F)(O)O | |
| 175.901 | |
| 175.9 |
Chemical Identifiers
| 143418-49-9 | |
| 175.901 | |
| UHDDEIOYXFXNNJ-UHFFFAOYSA-N | |
| 2734671 | |
| B(C1=CC(=C(C(=C1)F)F)F)(O)O |
| C6H4BF3O2 | |
| MFCD02093069 | |
| 3,4,5-trifluorophenyl boronic acid, 3,4,5-trifluorobenzeneboronic acid, 3,4,5-trifluorophenyl boranediol, boronic acid, b-3,4,5-trifluorophenyl, 3,4,5-trifluoro phenylboric acid, boronic acid, 3,4,5-trifluorophenyl, 3,4,5-trifluorophenylboronicacid, pubchem2340, acmc-209cqj | |
| (3,4,5-trifluorophenyl)boronic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P264b-P270-P280i-P301+P312-P305+P351+P338-P330-P501c
H302-H319
DOTInformation : Hazard Class: 9; Packaging Group: III
EINECSNumber : 000-000-0
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only