missing translation for 'onlineSavingsMsg'
Learn More

3,4,5-Trifluorobenzeneboronic acid, 97%

Catalog No. p-7047282
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL18519MD 250 mg
AAL1851903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL18519MD Supplier Thermo Scientific Chemicals Supplier No. L18519MD
Only null left

CAS: 143418-49-9 | C6H4BF3O2 | 175.901 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 143418-49-9
Molecular Formula C6H4BF3O2
Molecular Weight (g/mol) 175.901
MDL Number MFCD02093069
InChI Key UHDDEIOYXFXNNJ-UHFFFAOYSA-N
Synonym 3,4,5-trifluorophenyl boronic acid, 3,4,5-trifluorobenzeneboronic acid, 3,4,5-trifluorophenyl boranediol, boronic acid, b-3,4,5-trifluorophenyl, 3,4,5-trifluoro phenylboric acid, boronic acid, 3,4,5-trifluorophenyl, 3,4,5-trifluorophenylboronicacid, pubchem2340, acmc-209cqj
PubChem CID 2734671
IUPAC Name (3,4,5-trifluorophenyl)boronic acid
SMILES B(C1=CC(=C(C(=C1)F)F)F)(O)O

Specifications

Melting Point 275°C to 279°C
Quantity 250 mg
Beilstein 7371914
Formula Weight 175.9
Percent Purity 97%
Chemical Name or Material 3,4,5-Trifluorobenzeneboronic acid
Hazard Category H302-H319
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P264b-P270-P280i-P301+P312-P305+P351+P338-P330-P501c
DOTInformation Hazard Class: 9; Packaging Group: III
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only