missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,4,5-Trifluorobenzaldehyde 98.0+%, TCI America™
$51.98 - $150.16
Chemical Identifiers
| CAS | 132123-54-7 |
|---|---|
| Molecular Formula | C7H3F3O |
| Molecular Weight (g/mol) | 160.10 |
| MDL Number | MFCD00083523 |
| InChI Key | NLAVHUUABUFSIG-UHFFFAOYSA-N |
| Synonym | benzaldehyde, 3,4,5-trifluoro, 3,4,5-trifluoro-benzaldehyde, 3,4,5-ttrifluorobenzaldehyde, acmc-209bon, intermediates-zcf02014, 3,4,5-triflourobenzaldehyde, 3,4,5-trifluorobrnzaldehyde, ksc182i0f, 3,4,5-trifluorobenzaldehyde, benzaldehyde, 3,4,5-trifluoro-9ci |
| PubChem CID | 2776954 |
| IUPAC Name | 3,4,5-trifluorobenzaldehyde |
| SMILES | FC1=CC(C=O)=CC(F)=C1F |
Chemical Identifiers
| 132123-54-7 | |
| 160.10 | |
| NLAVHUUABUFSIG-UHFFFAOYSA-N | |
| 2776954 | |
| FC1=CC(C=O)=CC(F)=C1F |
| C7H3F3O | |
| MFCD00083523 | |
| benzaldehyde, 3,4,5-trifluoro, 3,4,5-trifluoro-benzaldehyde, 3,4,5-ttrifluorobenzaldehyde, acmc-209bon, intermediates-zcf02014, 3,4,5-triflourobenzaldehyde, 3,4,5-trifluorobrnzaldehyde, ksc182i0f, 3,4,5-trifluorobenzaldehyde, benzaldehyde, 3,4,5-trifluoro-9ci | |
| 3,4,5-trifluorobenzaldehyde |
Specifications
| 132123-54-7 | |
| 174°C | |
| MFCD00083523 | |
| benzaldehyde, 3,4,5-trifluoro, 3,4,5-trifluoro-benzaldehyde, 3,4,5-ttrifluorobenzaldehyde, acmc-209bon, intermediates-zcf02014, 3,4,5-triflourobenzaldehyde, 3,4,5-trifluorobrnzaldehyde, ksc182i0f, 3,4,5-trifluorobenzaldehyde, benzaldehyde, 3,4,5-trifluoro-9ci | |
| FC1=CC(C=O)=CC(F)=C1F | |
| 160.10 | |
| 160.10 | |
| Crystalline Lumps |
| White-Yellow | |
| C7H3F3O | |
| 1 g | |
| NLAVHUUABUFSIG-UHFFFAOYSA-N | |
| 3,4,5-trifluorobenzaldehyde | |
| 2776954 | |
| ≥98.0% (GC) | |
| 3,4,5-Trifluorobenzaldehyde |
Safety and Handling
TSCA : No