accessibility menu, dialog, popup

UserName

CAS RN 132123-54-7

FOFF

CAS RN 132123-54-7

IUPAC Name: 3,4,5-trifluorobenzaldehyde
Synonyms: 3,4,5-trifluorobenzaldehyde
Molecular Weight (g/mol): 160.1
Molecular Formula: C7H3F3O
InChi Key: NLAVHUUABUFSIG-UHFFFAOYSA-N
SMILES: FC1=CC(C=O)=CC(F)=C1F
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results