missing translation for 'onlineSavingsMsg'
Learn More

3,4,5-Trifluorobenzaldehyde 98.0+%, TCI America™

Catalog No. T23215G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T23215G 5 g
T23211G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T23215G Supplier TCI America Supplier No. T23215G
Only null left

Chemical Identifiers

CAS 132123-54-7
Molecular Formula C7H3F3O
Molecular Weight (g/mol) 160.10
MDL Number MFCD00083523
InChI Key NLAVHUUABUFSIG-UHFFFAOYSA-N
Synonym benzaldehyde, 3,4,5-trifluoro, 3,4,5-trifluoro-benzaldehyde, 3,4,5-ttrifluorobenzaldehyde, acmc-209bon, intermediates-zcf02014, 3,4,5-triflourobenzaldehyde, 3,4,5-trifluorobrnzaldehyde, ksc182i0f, 3,4,5-trifluorobenzaldehyde, benzaldehyde, 3,4,5-trifluoro-9ci
PubChem CID 2776954
IUPAC Name 3,4,5-trifluorobenzaldehyde
SMILES FC1=CC(C=O)=CC(F)=C1F

Specifications

Color White-Yellow
Boiling Point 174°C
Quantity 5 g
Formula Weight 160.10
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 3,4,5-Trifluorobenzaldehyde