missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,4,5-Trifluorobenzaldehyde 98.0+%, TCI America™
Specifications
Specifications
| Color | White-Yellow |
| Boiling Point | 174°C |
| Quantity | 1 g |
| Formula Weight | 160.10 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Lumps |
| Chemical Name or Material | 3,4,5-Trifluorobenzaldehyde |
Safety and Handling
| TSCA | No |