missing translation for 'onlineSavingsMsg'
Learn More

3,4,5-Trifluoroaniline 98.0+%, TCI America™

Catalog No. T23555G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T23555G 5 g
T23551G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T23555G Supplier TCI America Supplier No. T23555G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 163733-96-8
Molecular Formula C6H4F3N
Molecular Weight (g/mol) 147.10
MDL Number MFCD00083522
InChI Key SZRDJHHKIJHJHQ-UHFFFAOYSA-N
Synonym 3,4,5-trifluorobenzenamine, 3,4,5-trifluoro-phenylamine, benzenamine, 3,4,5-trifluoro, benzenamine, 3,4,5-trifluoro-9ci, 3,4,5-trifluorophenylamine, pubchem2298, 3,4,5-trifluoroanili, 3,4,5-trifluoraniline, 3,4,5-trifluroaniline, intermediates-zcf02231
PubChem CID 2724991
IUPAC Name 3,4,5-trifluoroaniline
SMILES NC1=CC(F)=C(F)C(F)=C1

Specifications

Melting Point 64°C
Color White-Yellow
Quantity 5 g
Formula Weight 147.10
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,4,5-Trifluoroaniline