accessibility menu, dialog, popup

UserName

CAS RN 163733-96-8

NH2FFF

CAS RN 163733-96-8

IUPAC Name: 3,4,5-trifluoroaniline
Synonyms: 3,4,5-trifluoroaniline
Molecular Weight (g/mol): 147.1
Molecular Formula: C6H4F3N
InChi Key: SZRDJHHKIJHJHQ-UHFFFAOYSA-N
SMILES: NC1=CC(F)=C(F)C(F)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results