missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,4,5-Tribromopyridine 98.0+%, TCI America™
Supplier: TCI America T32481G
Specifications
| 3,4,5-Tribromopyridine | |
| 107°C | |
| 285°C | |
| MFCD01646053 | |
| pubchem13500, 3,4,5-tribromo-pyridine, acmc-20a7i7, pyridine,3,4,5-tribromo, 3,4,5-tribromopyridine | |
| BrC1=CN=CC(Br)=C1Br | |
| 315.79 | |
| 315.79 | |
| Crystalline Powder |
| 2457-48-9 | |
| White-Yellow | |
| C5H2Br3N | |
| 1 g | |
| CWYVUHWKGWQPLP-UHFFFAOYSA-N | |
| 3,4,5-tribromopyridine | |
| 541462 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 2457-48-9 | |
| 315.79 | |
| CWYVUHWKGWQPLP-UHFFFAOYSA-N | |
| 541462 | |
| BrC1=CN=CC(Br)=C1Br |
| C5H2Br3N | |
| MFCD01646053 | |
| pubchem13500, 3,4,5-tribromo-pyridine, acmc-20a7i7, pyridine,3,4,5-tribromo, 3,4,5-tribromopyridine | |
| 3,4,5-tribromopyridine |
Safety and Handling
TSCA : No