missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,4,5-Tribromopyridine 98.0+%, TCI America™
$76.43 - $253.51
Chemical Identifiers
| CAS | 2457-48-9 |
|---|---|
| Molecular Formula | C5H2Br3N |
| Molecular Weight (g/mol) | 315.79 |
| MDL Number | MFCD01646053 |
| InChI Key | CWYVUHWKGWQPLP-UHFFFAOYSA-N |
| Synonym | pubchem13500, 3,4,5-tribromo-pyridine, acmc-20a7i7, pyridine,3,4,5-tribromo, 3,4,5-tribromopyridine |
| PubChem CID | 541462 |
| IUPAC Name | 3,4,5-tribromopyridine |
| SMILES | BrC1=CN=CC(Br)=C1Br |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
T3248200MG
|
TCI America
T3248200MG |
200 mg |
Each for $76.43
|
|
|||||
|
T32481G
|
TCI America
T32481G |
1 g |
Each for $253.51
|
|
|||||
Chemical Identifiers
| 2457-48-9 | |
| 315.79 | |
| CWYVUHWKGWQPLP-UHFFFAOYSA-N | |
| 541462 | |
| BrC1=CN=CC(Br)=C1Br |
| C5H2Br3N | |
| MFCD01646053 | |
| pubchem13500, 3,4,5-tribromo-pyridine, acmc-20a7i7, pyridine,3,4,5-tribromo, 3,4,5-tribromopyridine | |
| 3,4,5-tribromopyridine |
Specifications
| 2457-48-9 | |
| White-Yellow | |
| C5H2Br3N | |
| 200 mg | |
| CWYVUHWKGWQPLP-UHFFFAOYSA-N | |
| 3,4,5-tribromopyridine | |
| 541462 | |
| ≥98.0% (GC) | |
| 3,4,5-Tribromopyridine |
| 107°C | |
| 285°C | |
| MFCD01646053 | |
| pubchem13500, 3,4,5-tribromo-pyridine, acmc-20a7i7, pyridine,3,4,5-tribromo, 3,4,5-tribromopyridine | |
| BrC1=CN=CC(Br)=C1Br | |
| 315.79 | |
| 315.79 | |
| Crystalline Powder |
Safety and Handling
TSCA : No