missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,4,5-Tribromopyrazole 98.0+%, TCI America™
Supplier: TCI America T277725G
Specifications
| 3,4,5-Tribromopyrazole | |
| 188°C | |
| C3HBr3N2 | |
| 25 g | |
| TXQKCKQJBGFUBF-UHFFFAOYSA-N | |
| 3,4,5-tribromo-1H-pyrazole | |
| 627674 | |
| ≥98.0% (GC) |
| 17635-44-8 | |
| White-Yellow | |
| MFCD00040248 | |
| 3,4,5-tribromopyrazole, 1h-pyrazole, 3,4,5-tribromo, pyrazole, 3,4,5-tribromo, acmc-1c9yo, 3,4,5-tribromo pyrazole, ksc497i1h, #, 3,4,5-tribromo-1h-pyrazole 5g | |
| C1(=C(NN=C1Br)Br)Br | |
| 304.767 | |
| 304.77 | |
| Crystalline Powder |
Chemical Identifiers
| 17635-44-8 | |
| 304.767 | |
| TXQKCKQJBGFUBF-UHFFFAOYSA-N | |
| 627674 | |
| C1(=C(NN=C1Br)Br)Br |
| C3HBr3N2 | |
| MFCD00040248 | |
| 3,4,5-tribromopyrazole, 1h-pyrazole, 3,4,5-tribromo, pyrazole, 3,4,5-tribromo, acmc-1c9yo, 3,4,5-tribromo pyrazole, ksc497i1h, #, 3,4,5-tribromo-1h-pyrazole 5g | |
| 3,4,5-tribromo-1H-pyrazole |
Safety and Handling
TSCA : No